siRNA Details
| Gene Symbol: Tex22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029381.1 | NP_083657.1 | 3 | 1139 | ||
| GenBank mRNA | AF522069.1 | - | 3 | 1139 | ||
| AK006471.1 | - | 3 | 607 | |||
| BC048634.1 | - | 3 | 572 | 6770610 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex22| Gene Symbol: Tex22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029381.1 | NP_083657.1 | 3 | 1139 | ||
| GenBank mRNA | AF522069.1 | - | 3 | 1139 | ||
| AK006471.1 | - | 3 | 607 | |||
| BC048634.1 | - | 3 | 572 | 6770610 | ||