siRNA Details
| Gene Symbol: Clec4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029465.3 | NP_083741.1 | 9 | 827 | ||
| GenBank mRNA | AI449163.1 | - | 1 | 409 | ||
| AK079992.1 | - | 9 | 828 | |||
| BC025069.1 | - | 9 | 819 | |||
| BC087963.1 | - | 9 | 827 | 6766377 | ||

Clec4g
Genomic Map