siRNA Details
| Gene Symbol: Clec4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029465.3 | NP_083741.1 | 5 | 348 | ||
| GenBank mRNA | AK079992.1 | - | 5 | 349 | ||
| BC025069.1 | - | 5 | 340 | |||
| BC087963.1 | - | 5 | 348 | 6766377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec4g| Gene Symbol: Clec4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029465.3 | NP_083741.1 | 5 | 348 | ||
| GenBank mRNA | AK079992.1 | - | 5 | 349 | ||
| BC025069.1 | - | 5 | 340 | |||
| BC087963.1 | - | 5 | 348 | 6766377 | ||