siRNA Details
| Gene Symbol: Ift43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199843.1 | NP_001186772.1 | 6, 7 | 385 | ||
| NM_029601.3 | NP_083877.1 | 5, 6 | 352 | |||
| GenBank mRNA | AK006115.1 | - | 5, 6 | 350 | ||
| BC034668.1 | - | 6, 7 | 323 | |||
| BC038915.1 | - | 6, 7 | 374 | 5148723 | ||
| BE987571.1 | - | 3, 4 | 437 | |||
| CF751010.1 | - | 6, 7 | 348 | |||

Ift43
Genomic Map