siRNA Details
Gene Symbol: Ift43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199843.1 | NP_001186772.1 | 9 | 626 | ||
NM_029601.3 | NP_083877.1 | 8 | 593 | |||
GenBank mRNA | AK006115.1 | - | 8 | 591 | ||
BC034668.1 | - | 9 | 564 | |||
BC038915.1 | - | 9 | 615 | 5148723 | ||
BE987571.1 | - | 1 | 196 | |||
CF751010.1 | - | 9 | 589 |
