siRNA Details
Gene Symbol: Ddrgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029832.2 | NP_084108.1 | 8, 9 | 860 | ||
GenBank mRNA | BC051541.1 | - | 6, 7 | 449 | ||
BC115616.2 | - | 8, 9 | 860 | 40102754 | ||
BC115617.1 | - | 8, 9 | 860 | 40102757 |
