siRNA Details
| Gene Symbol: 2510039O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029841.3 | NP_084117.2 | 2 | 1221 | ||
| GenBank mRNA | AK011059.1 | - | 2 | 1221 | ||
| AK052853.1 | - | 1 | 1592 | |||
| AK153761.1 | - | 2 | 1222 | |||
| AK173332.1 | - | 3 | 1076 | |||
| BC012878.1 | - | 2 | 1194 | 4190292 | ||

2510039O18Rik
Genomic Map