siRNA Details
Gene Symbol: 2200002D01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033738.1 | - | 632 |
Gene Symbol: Yif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110201.1 | NP_001103671.1 | 8 | 920 | ||
NM_029887.3 | NP_084163.2 | 8 | 929 | |||
GenBank mRNA | AK020444.1 | - | 8 | 900 | ||
AK135529.1 | - | 898 | ||||
AW743704.1 | - | 1 | 212 | |||
BC109331.2 | - | 8 | 899 | 40051521 |
