siRNA Details
| Gene Symbol: Cstad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030137.2 | NP_084413.1 | 2 | 94 | ||
| GenBank mRNA | AK020589.1 | - | 2 | 96 | ||
| AK047294.1 | - | 2 | 95 | |||
| BC117017.1 | - | 2 | 90 | 40126336 | ||
| BC117019.1 | - | 2 | 90 | 40126338 | ||

Cstad
Genomic Map