siRNA Details
| Gene Symbol: Gpr146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038703.2 | NP_001033792.1 | 2 | 739 | ||
| NM_030258.4 | NP_084534.2 | 2 | 664 | |||
| GenBank mRNA | AK046512.1 | - | 2 | 687 | ||
| AK050034.1 | - | 2 | 633 | |||
| AK087100.1 | - | 2 | 633 | |||
| AK137454.1 | - | 2 | 741 | |||
| AK154708.1 | - | 2 | 685 | |||
| AK155876.1 | - | 2 | 667 | |||
More Information
Set Membership: |
|

Gpr146
Genomic Map