siRNA Details
| Gene Symbol: Ngrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031375.4 | NP_113552.3 | 3 | 737 | ||
| GenBank mRNA | AB047544.1 | - | 3 | 763 | ||
| AK083330.1 | - | 2 | 1130 | |||
| AK144756.1 | - | 3 | 733 | |||
| BC004034.1 | - | 724 | 3495639 | |||

Ngrn
Genomic Map