siRNA Details
| Gene Symbol: Cml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053096.3 | NP_444326.2 | 3 | 1000 | ||
| GenBank mRNA | AF163315.1 | - | 3 | 978 | ||
| AK050449.1 | - | 3 | 930 | |||
| BC039773.1 | - | 3 | 1020 | 4193874 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cml2| Gene Symbol: Cml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053096.3 | NP_444326.2 | 3 | 1000 | ||
| GenBank mRNA | AF163315.1 | - | 3 | 978 | ||
| AK050449.1 | - | 3 | 930 | |||
| BC039773.1 | - | 3 | 1020 | 4193874 | ||