siRNA Details
| Gene Symbol: Wwtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168281.1 | NP_001161753.1 | 2, 3 | 624 | ||
| NM_133784.3 | NP_598545.2 | 2, 3 | 703 | |||
| GenBank mRNA | AJ299430.1 | - | 2, 3 | 638 | ||
| AK009983.1 | - | 2, 3 | 636 | |||
| AK011504.1 | - | 2, 3 | 679 | |||
| AK142299.1 | - | 2, 3 | 625 | |||
| AK142720.1 | - | 2, 3 | 628 | |||
| BC004640.1 | - | 2, 3 | 624 | 3157246 | ||
| BC014727.1 | - | 2, 3 | 655 | 4487316 | ||

Wwtr1
Genomic Map