siRNA Details
| Gene Symbol: Oog3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201258.2 | NP_957710.2 | 3 | 412 | ||
| GenBank mRNA | AK136030.1 | - | 3 | 413 | ||
| AY573563.1 | - | 3 | 408 | |||
| BC053700.1 | - | 3 | 408 | 30049894 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oog3| Gene Symbol: Oog3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201258.2 | NP_957710.2 | 3 | 412 | ||
| GenBank mRNA | AK136030.1 | - | 3 | 413 | ||
| AY573563.1 | - | 3 | 408 | |||
| BC053700.1 | - | 3 | 408 | 30049894 | ||