siRNA Details
| Gene Symbol: Wfdc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081957.1 | NP_001075426.1 | 1 | 92 | ||
| GenBank mRNA | BC038870.1 | - | 1 | 21 | ||
| BC089618.1 | - | 1 | 65 | 6766535 | ||
| FJ007372.1 | - | 1 | 133 | |||
| W08116.1 | - | 1 | 92 | |||

Wfdc17
Genomic Map