siRNA Details
| Gene Symbol: Dnajc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008929.3 | NP_032955.2 | 7, 8 | 996 | ||
| GenBank mRNA | AK004033.1 | - | 2, 3 | 297 | ||
| AK146097.1 | - | 7, 8 | 960 | |||
| AK148270.1 | - | 7, 8 | 953 | |||
| AK188318.1 | - | 1, 2 | 71 | |||
| BC013766.1 | - | 7, 8 | 941 | 2646147 | ||
| U28423.1 | - | 7, 8 | 939 | |||

Dnajc3
Genomic Map