siRNA Details
| Gene Symbol: Dnajc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008929.3 | NP_032955.2 | 8 | 1031 | ||
| GenBank mRNA | AK004033.1 | - | 3 | 332 | ||
| AK146097.1 | - | 8 | 995 | |||
| AK148270.1 | - | 8 | 988 | |||
| AK188318.1 | - | 2 | 106 | |||
| BC013766.1 | - | 8 | 976 | 2646147 | ||
| U28423.1 | - | 8 | 974 | |||

Dnajc3
Genomic Map