siRNA Details
Gene Symbol: Fam129c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166213.1 | NP_001159685.1 | 3, 4 | 423 | ||
GenBank mRNA | AK158308.1 | - | 3, 4 | 288 |
Gene Symbol: Pgls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036059.1 | - | 918 |
