siRNA Details
| Gene Symbol: Fam129c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166213.1 | NP_001159685.1 | 4 | 498 | ||
| GenBank mRNA | AK158308.1 | - | 4 | 363 | ||
| Gene Symbol: Pgls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036059.1 | - | 993 | |||

Fam129c
Genomic Map