siRNA Details
| Gene Symbol: Gm11837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243100.1 | NP_001230029.1 | 3 | 249 | ||
| GenBank mRNA | AK005641.1 | - | 2 | 36 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11837| Gene Symbol: Gm11837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243100.1 | NP_001230029.1 | 3 | 249 | ||
| GenBank mRNA | AK005641.1 | - | 2 | 36 | ||