siRNA Details
Gene Symbol: Gm11837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243100.1 | NP_001230029.1 | 5 | 406 | ||
GenBank mRNA | AK005641.1 | - | 5 | 194 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm11837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243100.1 | NP_001230029.1 | 5 | 406 | ||
GenBank mRNA | AK005641.1 | - | 5 | 194 |