siRNA Details
| Gene Symbol: Gm1971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143472.1 | - | 1 | 1535 | ||
| AK165166.1 | - | 1 | 1535 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm1971| Gene Symbol: Gm1971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143472.1 | - | 1 | 1535 | ||
| AK165166.1 | - | 1 | 1535 | |||