siRNA Details
| Gene Symbol: Hrct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027511.1 | NP_081787.1 | 1 | 239 | ||
| GenBank mRNA | AK014425.1 | - | 240 | |||
| BC080752.1 | - | 1 | 131 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hrct1| Gene Symbol: Hrct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027511.1 | NP_081787.1 | 1 | 239 | ||
| GenBank mRNA | AK014425.1 | - | 240 | |||
| BC080752.1 | - | 1 | 131 | |||