siRNA Details
| Gene Symbol: Fancf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115087.1 | NP_001108559.1 | 1 | 907 | ||
| GenBank mRNA | AK139575.1 | - | 1 | 378 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fancf| Gene Symbol: Fancf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115087.1 | NP_001108559.1 | 1 | 907 | ||
| GenBank mRNA | AK139575.1 | - | 1 | 378 | ||