siRNA Details
| Gene Symbol: Gm3519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139157.1 | - | 1, 2 | 499 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm3519| Gene Symbol: Gm3519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139157.1 | - | 1, 2 | 499 | ||