siRNA Details
Gene Symbol: 9130023H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177001.3 | NP_795975.1 | 1 | 1419 | ||
GenBank mRNA | AK033604.1 | - | 1 | 1379 | ||
AK084086.1 | - | 1 | 1422 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9130023H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177001.3 | NP_795975.1 | 1 | 1419 | ||
GenBank mRNA | AK033604.1 | - | 1 | 1379 | ||
AK084086.1 | - | 1 | 1422 |