siRNA Details
| Gene Symbol: LOC100043315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC106998.1 | - | 2, 3 | 406 | ||
| BC106999.1 | - | 2, 3 | 315 | |||
| BC114343.1 | - | 2, 3 | 315 | |||
| BC115945.1 | - | 2, 3 | 315 | |||
| BC115946.1 | - | 2, 3 | 315 | |||
| BC127134.1 | - | 2, 3 | 315 | |||

LOC100043315
Genomic Map