siRNA Details
Gene Symbol: BC030499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287206.1 | NP_001274135.1 | 12 | 1139 | ||
GenBank mRNA | AK138440.1 | - | 9 | 1548 | ||
BC030499.1 | - | 3 | 387 | |||
BC059191.1 | - | 11 | 1445 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008768145, XM_008768146, XM_008768147, XM_006246951.
Panther Classification:
Gene Ontology Categories:
