siRNA Details
| Gene Symbol: Mipep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC059092.1 | - | 2 | 662 | ||
| BC144798.1 | - | 20 | 3345 | |||
| BC145711.1 | - | 20 | 3345 | 40131014 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mipep| Gene Symbol: Mipep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC059092.1 | - | 2 | 662 | ||
| BC144798.1 | - | 20 | 3345 | |||
| BC145711.1 | - | 20 | 3345 | 40131014 | ||