siRNA Details
| Gene Symbol: Srcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK154145.1 | - | 12, 13 | 1763 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srcap| Gene Symbol: Srcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK154145.1 | - | 12, 13 | 1763 | ||