siRNA Details
| Gene Symbol: Map3k9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174107.1 | NP_001167578.1 | 10 | 2261 | ||
| GenBank mRNA | AK155677.1 | - | 10 | 2267 | ||
| BC030944.1 | - | 7 | 930 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k9| Gene Symbol: Map3k9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174107.1 | NP_001167578.1 | 10 | 2261 | ||
| GenBank mRNA | AK155677.1 | - | 10 | 2267 | ||
| BC030944.1 | - | 7 | 930 | |||