siRNA Details
| Gene Symbol: Emx2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK033011.1 | - | 1 | 67 | ||
| AY117414.1 | - | LIT1111 EMX2OS |
1 | 418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emx2os| Gene Symbol: Emx2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AK033011.1 | - | 1 | 67 | ||
| AY117414.1 | - | LIT1111 EMX2OS |
1 | 418 | ||