siRNA Details
| Gene Symbol: 2210409E12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008869.1 | - | 556 | |||
| Gene Symbol: Gm10367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK147092.1 | - | 1 | 1510 | ||

2210409E12Rik
Genomic Map