siRNA Details
| Gene Symbol: Zfp951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BY735223.1 | - | 311 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp951| Gene Symbol: Zfp951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BY735223.1 | - | 311 | |||