siRNA Details
| Gene Symbol: Srrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK171001.1 | - | 2 | 294 | ||
| BC019803.1 | - | 2 | 280 | |||
| BC037186.1 | - | 2 | 238 | |||
| BC044058.1 | - | 1 | 65 | |||
| BC092355.1 | - | 2 | 158 | |||
| BY202609.1 | - | 2 | 294 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srrm2| Gene Symbol: Srrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK171001.1 | - | 2 | 294 | ||
| BC019803.1 | - | 2 | 280 | |||
| BC037186.1 | - | 2 | 238 | |||
| BC044058.1 | - | 1 | 65 | |||
| BC092355.1 | - | 2 | 158 | |||
| BY202609.1 | - | 2 | 294 | |||