siRNA Details
| Gene Symbol: Srrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK171001.1 | - | 2 | 456 | ||
| BC019803.1 | - | 2 | 442 | |||
| BC037186.1 | - | 2 | 400 | |||
| BC044058.1 | - | 1 | 227 | |||
| BC092355.1 | - | 2 | 320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srrm2| Gene Symbol: Srrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK171001.1 | - | 2 | 456 | ||
| BC019803.1 | - | 2 | 442 | |||
| BC037186.1 | - | 2 | 400 | |||
| BC044058.1 | - | 1 | 227 | |||
| BC092355.1 | - | 2 | 320 | |||