siRNA Details
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197024.1 | NP_001183953.1 | 8 | 1021 | ||
| NM_001290736.1 | NP_001277665.1 | 8 | 1032 | |||
| GenBank mRNA | BC059910.1 | - | 8 | 1032 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unkl| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197024.1 | NP_001183953.1 | 8 | 1021 | ||
| NM_001290736.1 | NP_001277665.1 | 8 | 1032 | |||
| GenBank mRNA | BC059910.1 | - | 8 | 1032 | ||