siRNA Details
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197024.1 | NP_001183953.1 | 12 | 1572 | ||
| NM_001290736.1 | NP_001277665.1 | 12 | 1583 | |||
| GenBank mRNA | BC059910.1 | - | 12 | 1583 | ||
| BC089378.1 | - | 3 | 325 | |||

Unkl
Genomic Map