siRNA Details
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197024.1 | NP_001183953.1 | 2 | 178 | ||
| NM_001290736.1 | NP_001277665.1 | 2 | 189 | |||
| GenBank mRNA | AK054549.1 | - | 1 | 1163 | ||
| BC059910.1 | - | 2 | 189 | |||
| CJ068427.1 | - | 2 | 178 | |||

Unkl
Genomic Map