siRNA Details
| Gene Symbol: Cfap61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC024760.1 | - | 1223 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap61| Gene Symbol: Cfap61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC024760.1 | - | 1223 | |||