siRNA Details
| Gene Symbol: C230062I16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082547.1 | - | 78 | |||
| Gene Symbol: Zfp74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051279.1 | - | 1 | 366 | ||

C230062I16Rik
Genomic Map