siRNA Details
| Gene Symbol: Phactr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161797.1 | NP_001155269.1 | 7 | 1501 | ||
| NM_175306.4 | NP_780515.2 | 8 | 1582 | |||
| GenBank mRNA | AK049209.1 | - | 8 | 1545 | ||
| AK220496.1 | - | 7 | 1480 | |||
| BC075672.1 | - | 8 | 1544 | 30549496 | ||
| BC096033.1 | - | 8 | 1501 | 6854767 | ||

Phactr4
Genomic Map