siRNA Details
| Gene Symbol: Gpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133884.2 | NP_598645.2 | 1 | 457 | ||
| GenBank mRNA | BC018407.1 | - | 1 | 450 | 4015340 | |
| CN721622.1 | - | 1 | 59 | |||
| CO798620.2 | - | 1 | 457 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpn2| Gene Symbol: Gpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133884.2 | NP_598645.2 | 1 | 457 | ||
| GenBank mRNA | BC018407.1 | - | 1 | 450 | 4015340 | |
| CN721622.1 | - | 1 | 59 | |||
| CO798620.2 | - | 1 | 457 | |||