siRNA Details
| Gene Symbol: N4bp2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133898.4 | NP_598659.3 | 1 | 203 | ||
| GenBank mRNA | AK010587.1 | - | 1 | 207 | ||
| AK042145.1 | - | 1 | 179 | |||
| AK046116.1 | - | 1 | 179 | |||
| AK131647.1 | - | 1 | 195 | |||
| AK138666.1 | - | 1 | 179 | |||
| AK155341.1 | - | 1 | 179 | |||
| BB629746.1 | - | 1 | 203 | |||

N4bp2l1
Genomic Map