siRNA Details
| Gene Symbol: N4bp2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133898.4 | NP_598659.3 | 5 | 792 | ||
| GenBank mRNA | AK046116.1 | - | 5 | 769 | ||
| AK131647.1 | - | 5 | 784 | |||
| BC019754.1 | - | 4 | 754 | 4007588 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

N4bp2l1| Gene Symbol: N4bp2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133898.4 | NP_598659.3 | 5 | 792 | ||
| GenBank mRNA | AK046116.1 | - | 5 | 769 | ||
| AK131647.1 | - | 5 | 784 | |||
| BC019754.1 | - | 4 | 754 | 4007588 | ||