siRNA Details
| Gene Symbol: Hscb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153571.2 | NP_705799.2 | 3 | 418 | ||
| GenBank mRNA | AK131961.1 | - | 3 | 420 | ||
| BC027641.1 | - | 3 | 382 | 1244839 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hscb| Gene Symbol: Hscb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153571.2 | NP_705799.2 | 3 | 418 | ||
| GenBank mRNA | AK131961.1 | - | 3 | 420 | ||
| BC027641.1 | - | 3 | 382 | 1244839 | ||