siRNA Details
Gene Symbol: Hscb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153571.2 | NP_705799.2 | 3, 4 | 457 | ||
GenBank mRNA | AK131961.1 | - | 3, 4 | 459 | ||
BC027641.1 | - | 3, 4 | 421 | 1244839 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hscb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153571.2 | NP_705799.2 | 3, 4 | 457 | ||
GenBank mRNA | AK131961.1 | - | 3, 4 | 459 | ||
BC027641.1 | - | 3, 4 | 421 | 1244839 |