siRNA Details
Gene Symbol: Nfxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133921.2 | NP_598682.2 | 7 | 1031 | ||
GenBank mRNA | AK132320.1 | - | 7 | 1032 | ||
AK162215.1 | - | 6 | 1304 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nfxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133921.2 | NP_598682.2 | 7 | 1031 | ||
GenBank mRNA | AK132320.1 | - | 7 | 1032 | ||
AK162215.1 | - | 6 | 1304 |