siRNA Details
| Gene Symbol: Zfp282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146175.3 | NP_666287.3 | 7, 8 | 1305 | ||
| GenBank mRNA | AK030516.1 | - | 7, 8 | 1213 | ||
| AK088531.1 | - | 7, 8 | 1304 | |||
| AK148038.1 | - | 7, 8 | 1291 | |||
| AK185073.1 | - | 51 | ||||
| AK186362.1 | - | 51 | ||||
| BC025581.1 | - | 7, 8 | 1207 | 5325029 | ||

Zfp282
Genomic Map