siRNA Details
| Gene Symbol: Zfp282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146175.3 | NP_666287.3 | 2 | 468 | ||
| GenBank mRNA | AK030516.1 | - | 2 | 376 | ||
| AK081854.1 | - | 2 | 367 | |||
| AK088531.1 | - | 2 | 467 | |||
| AK148038.1 | - | 2 | 454 | |||
| BC025581.1 | - | 2 | 370 | 5325029 | ||

Zfp282
Genomic Map